2,2,2-Trifluoro-1-(4-(trifluoromethoxy)-[1,1'-biphenyl]-2-yl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
64331-1G
64331-500MG
64331-5G
64331-25G
77472-384EA
415.09
USD
77472-384
77485-010
77495-566
77504-876
2,2,2-Trifluoro-1-(4-(trifluoromethoxy)-[1,1'-biphenyl]-2-yl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-(trifluoromethoxy)-[1,1'-biphenyl]-2-yl)ethanone
Caution: For research use only
Formula:
C₁₅H₈F₆O₂ Storage Temperature: Ambient |
MDL Number:
MFCD34814296 CAS Number: 2921875-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...