(2-(Cyclopropylmethoxy)-3,5-dimethylphenyl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
356289-1G
356289-500MG
356289-5G
356289-25G
77470-220EA
415.09
USD
77470-220
77482-888
77493-322
77503-108
(2-(Cyclopropylmethoxy)-3,5-dimethylphenyl)(methyl)sulfane ≥95%
(2-(Cyclopropylmethoxy)-3,5-dimethylphenyl)(methyl)sulfane
Caution: For research use only
Formula:
C₁₃H₁₈OS Storage Temperature: Ambient |
CAS Number:
3002422-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...