4'-Benzyloxy-3'-fluoroacetophenone ≥96%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-(4-(Benzyloxy)-3-fluorophenyl)ethanone
273497-5G
273497-500MG
273497-1G
273497-25G
77490-972EA
297
USD
77490-972
77480-624
77467-928
4'-Benzyloxy-3'-fluoroacetophenone ≥96%
4'-Benzyloxy-3'-fluoroacetophenone
Caution: For research use only
Formula:
C₁₅H₁₃FO₂ MW: 244.261 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659550 CAS Number: 81227-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...