2-Bromo-5-(bromomethyl)-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

20539-1G 20539-500MG 20539-5G
77466-688EA 457.32 USD
77466-688 77479-420 77489-692
2-Bromo-5-(bromomethyl)-1,3-dimethylbenzene ≥95%
2-Bromo-5-(bromomethyl)-1,3-dimethylbenzene

Caution: For research use only

Formula: C₉H₁₀Br₂
Storage Temperature: Ambient
MDL Number: MFCD18907460
CAS Number: 35510-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR