1-(2,5-Dichlorophenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥97%

Supplier: AOBChem USA

Warning

183490-1G 183490-500MG 183490-5G
77466-224EA 594.31 USD
77466-224 77478-968 77489-202
1-(2,5-Dichlorophenyl)-2,2,3,3,3-pentafluoro-1-propanone ≥97%
1-(2,5-Dichlorophenyl)-2,2,3,3,3-pentafluoro-1-propanone

Caution: For research use only

Formula: C₉H₃Cl₂F₅O
Storage Temperature: Ambient
MDL Number: MFCD16613959
CAS Number: 1916280-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR