(3-Ethylphenyl)(phenyl)methanol ≥95%
Supplier: AOBChem USA
|
Warning
|
139659-1G
139659-500MG
139659-5G
139659-25G
77464-764EA
397.58
USD
77464-764
77477-542
77487-658
77498-406
(3-Ethylphenyl)(phenyl)methanol ≥95%
(3-Ethylphenyl)(phenyl)methanol
Caution: For research use only
Formula:
C₁₅H₁₆O Storage Temperature: Ambient |
MDL Number:
MFCD24127229 CAS Number: 108125-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...