(2,6-Difluoropyridin-3-yl)(furan-2-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

137492-1G 137492-500MG 137492-5G 137492-25G
77464-708EA 415.09 USD
77464-708 77477-488 77487-602 77498-388
(2,6-Difluoropyridin-3-yl)(furan-2-yl)methanol ≥97%
(2,6-Difluoropyridin-3-yl)(furan-2-yl)methanol

Caution: For research use only

Formula: C₁₀H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD34832862
CAS Number: 2921894-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR