1-(3,5-dimethyl-2-((4-methylbenzyl)oxy)phenyl)propan-1-ol

Supplier: AOBChem USA

356213-500MG 356213-5G 356213-25G
77503-102EA 4812 USD
77503-102 77493-316 77470-214 77482-882
1-(3,5-dimethyl-2-((4-methylbenzyl)oxy)phenyl)propan-1-ol
1-(3,5-dimethyl-2-((4-methylbenzyl)oxy)phenyl)propan-1-ol

Caution: For research use only

Formula: C₁₉H₂₄O₂
MW: 284.393 g/mol
CAS Number: 3002516-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR