3,5-Dimethyl-2-((4-methylbenzyl)oxy)benzoic acid

Supplier: AOBChem USA

356198-500MG 356198-25G 356198-5G 356198-1G
77493-304EA 1441 USD
77493-304 77470-202 77503-090 77482-870
3,5-Dimethyl-2-((4-methylbenzyl)oxy)benzoic acid
3,5-Dimethyl-2-((4-methylbenzyl)oxy)benzoic acid

Caution: For research use only

Formula: C₁₇H₁₈O₃
MW: 270.323 g/mol
CAS Number: 3002442-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR