2-Bromo-6-fluoro-3-(trifluoromethyl)benzonitrile

Supplier: AOBChem USA

35459-500MG 35459-5G 35459-1G
77470-150EA 469 USD
77470-150 77482-818 77493-250
2-Bromo-6-fluoro-3-(trifluoromethyl)benzonitrile
2-Bromo-6-fluoro-3-(trifluoromethyl)benzonitrile

Caution: For research use only

Formula: C₈H₂BrF₄N
MW: 268.006 g/mol
MDL Number: MFCD31568568
CAS Number: 2091606-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR