Ethyl(4'-methyl-[1,1'-biphenyl]-4-yl)sulfane

Supplier: AOBChem USA

353375-5G 353375-500MG 353375-25G 353375-1G
77493-230EA 1441 USD
77493-230 77470-130 77482-798 77503-028
Ethyl(4'-methyl-[1,1'-biphenyl]-4-yl)sulfane
Ethyl(4'-methyl-[1,1'-biphenyl]-4-yl)sulfane

Caution: For research use only

Formula: C₁₅H₁₆S
MW: 228.353 g/mol
CAS Number: 3002487-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR