4-([1,1'-biphenyl]-3-yl)tetrahydro-2H-thiopyran-4-ol

Supplier: AOBChem USA

353290-1G 353290-5G 353290-25G 353290-500MG
77503-024EA 4812 USD
77503-024 77493-226 77470-126 77482-794
4-([1,1'-biphenyl]-3-yl)tetrahydro-2H-thiopyran-4-ol
4-([1,1'-biphenyl]-3-yl)tetrahydro-2H-thiopyran-4-ol

Caution: For research use only

Formula: C₁₇H₁₈OS
MW: 270.389 g/mol
CAS Number: 3002444-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR