(4-Isobutoxyphenyl)(methyl)sulfane
Supplier: AOBChem USA
347009-1G
347009-500MG
347009-25G
347009-5G
77482-718EA
273
USD
77482-718
77493-146
77470-050
77502-954
(4-Isobutoxyphenyl)(methyl)sulfane
(4-Isobutoxyphenyl)(methyl)sulfane
Caution: For research use only
Formula:
C₁₁H₁₆OS MW: 196.3091 g/mol |
CAS Number:
1443310-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...