1-(3-((2-fluorobenzyl)oxy)-5-methylphenyl)prop-2-en-1-ol

Supplier: AOBChem USA

346857-25G 346857-1G 346857-500MG 346857-5G
77493-142EA 1441 USD
77493-142 77502-950 77482-714 77470-046
1-(3-((2-fluorobenzyl)oxy)-5-methylphenyl)prop-2-en-1-ol
1-(3-((2-fluorobenzyl)oxy)-5-methylphenyl)prop-2-en-1-ol

Caution: For research use only

Formula: C₁₇H₁₇FO₂
MW: 272.314 g/mol
CAS Number: 3002443-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR