(6-Bromo-2-fluoro-3-methylphenyl)(p-tolyl)methanol

Supplier: AOBChem USA

346696-1G 346696-25G 346696-5G 346696-500MG
77482-702EA 288 USD
77482-702 77493-128 77502-936 77470-034
(6-Bromo-2-fluoro-3-methylphenyl)(p-tolyl)methanol
(6-Bromo-2-fluoro-3-methylphenyl)(p-tolyl)methanol

Caution: For research use only

Formula: C₁₅H₁₄BrFO
MW: 309.173 g/mol
CAS Number: 3002453-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR