1-(3-Bromobenzo[b]thiophen-2-yl)-2-methylpropan-1-ol

Supplier: AOBChem USA

346675-5G 346675-25G 346675-500MG 346675-1G
77482-690EA 288 USD
77482-690 77493-116 77502-924 77470-022
1-(3-Bromobenzo[b]thiophen-2-yl)-2-methylpropan-1-ol
1-(3-Bromobenzo[b]thiophen-2-yl)-2-methylpropan-1-ol

Caution: For research use only

Formula: C₁₂H₁₃BrOS
MW: 285.2 g/mol
CAS Number: 3002544-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR