(3-Bromobenzo[b]thiophen-2-yl)(3-chlorophenyl)methanol

Supplier: AOBChem USA

346673-500MG 346673-5G 346673-25G 346673-1G
77493-112EA 1441 USD
77493-112 77482-686 77470-018 77502-920
(3-Bromobenzo[b]thiophen-2-yl)(3-chlorophenyl)methanol
(3-Bromobenzo[b]thiophen-2-yl)(3-chlorophenyl)methanol

Caution: For research use only

Formula: C₁₅H₁₀BrClOS
MW: 353.661 g/mol
CAS Number: 3002504-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR