(6-Chloro-2-fluoro-3-methylphenyl)(4-methoxyphenyl)methanol

Supplier: AOBChem USA

346500-5G 346500-25G 346500-500MG 346500-1G
77493-068EA 1441 USD
77493-068 77469-974 77482-642 77502-876
(6-Chloro-2-fluoro-3-methylphenyl)(4-methoxyphenyl)methanol
(6-Chloro-2-fluoro-3-methylphenyl)(4-methoxyphenyl)methanol

Caution: For research use only

Formula: C₁₅H₁₄ClFO₂
MW: 280.722 g/mol
CAS Number: 3002543-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR