(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(cyclopropyl)methanol

Supplier: AOBChem USA

346270-5G 346270-1G 346270-500MG 346270-25G
77482-590EA 288 USD
77482-590 77469-922 77493-016 77502-824
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(cyclopropyl)methanol
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)(cyclopropyl)methanol

Caution: For research use only

Formula: C₁₆H₁₄BrFO
MW: 321.1841 g/mol
CAS Number: 3002476-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR