Butyl(3-ethylphenyl)sulfane ≥97%

Supplier: AOBChem USA

332682-1G 332682-500MG 332682-5G 332682-25G
77469-586EA 163.77 USD
77469-586 77482-260 77492-672 77502-520
Butyl(3-ethylphenyl)sulfane ≥97%
Butyl(3-ethylphenyl)sulfane

Caution: For research use only

Formula: C₁₂H₁₈S
CAS Number: 3002485-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR