(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(5-methylthiophen-2-yl)methanol ≥95%
Supplier: AOBChem USA
327124-1G
327124-500MG
327124-5G
327124-25G
77469-386EA
415.09
USD
77469-386
77482-062
77492-466
77502-342
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(5-methylthiophen-2-yl)methanol ≥95%
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(5-methylthiophen-2-yl)methanol
Caution: For research use only
Formula:
C₁₃H₉BrF₄OS |
CAS Number:
3002456-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...