3-Fluoro-2-hydroxy-5-methoxybenzaldehyde ≥97%
Supplier: AOBChem USA
32526-1G
32526-500MG
32526-5G
77469-326EA
1042.36
USD
77469-326
77482-002
77492-406
3-Fluoro-2-hydroxy-5-methoxybenzaldehyde ≥97%
3-Fluoro-2-hydroxy-5-methoxybenzaldehyde
Caution: For research use only
Formula:
C₈H₇FO₃ |
MDL Number:
MFCD23705523 CAS Number: 1417996-86-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...