1-(5-Bromo-2-fluoro-3-(methylthio)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

319921-1G 319921-500MG 319921-5G 319921-25G
77469-150EA 415.09 USD
77469-150 77481-826 77492-224 77502-156
1-(5-Bromo-2-fluoro-3-(methylthio)phenyl)propan-1-one ≥95%
1-(5-Bromo-2-fluoro-3-(methylthio)phenyl)propan-1-one

Caution: For research use only

Formula: C₁₀H₁₀BrFOS
CAS Number: 3002453-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR