3-Bromo-5-chloro-N,N-dimethylaniline ≥95%

Supplier: AOBChem USA

Warning

297677-1G 297677-500MG 297677-5G
77468-414EA 235.87 USD
77468-414 77481-100 77491-472
3-Bromo-5-chloro-N,N-dimethylaniline ≥95%
3-Bromo-5-chloro-N,N-dimethylaniline

Caution: For research use only

Formula: C₈H₉BrClN
Storage Temperature: Ambient
MDL Number: MFCD29092535
CAS Number: 1369856-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR