(2,3-Difluoro-4-methoxyphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

291363-1G 291363-500MG 291363-5G 291363-25G
77468-226EA 415.09 USD
77468-226 77480-912 77491-276 77501-288
(2,3-Difluoro-4-methoxyphenyl)triethylsilane ≥95%
(2,3-Difluoro-4-methoxyphenyl)triethylsilane

Caution: For research use only

Formula: C₁₃H₂₀F₂OSi
Storage Temperature: Ambient
MDL Number: MFCD34888096
CAS Number: 2969473-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR