3-Chloro-4-(methylsulfanyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

24356-1G 24356-500MG 24356-5G 24356-25G
77467-326EA 195.7 USD
77467-326 77480-050 77490-358 77500-518
3-Chloro-4-(methylsulfanyl)benzoic acid ≥97%
3-Chloro-4-(methylsulfanyl)benzoic acid

Caution: For research use only

Formula: C₈H₇ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD17291669
CAS Number: 121120-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR