α-Butyl-4-(methylthio)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

214599-1G 214599-500MG 214599-5G 214599-25G
77466-926EA 397.58 USD
77466-926 77479-658 77489-944 77500-200
α-Butyl-4-(methylthio)benzenemethanol ≥95%
α-Butyl-4-(methylthio)benzenemethanol

Caution: For research use only

Formula: C₁₂H₁₈OS
Storage Temperature: Ambient
MDL Number: MFCD34867696
CAS Number: 1603406-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR