Ethyl-5-fluoronicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 5-fluoronicotinate

20253-1G 20253-500MG 20253-5G 20253-25G
77466-614EA 111.24 USD
77466-614 77479-348 77489-614 77499-934
Ethyl-5-fluoronicotinate ≥97%
Ethyl-5-fluoronicotinate

Caution: For research use only

Formula: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD01692096
CAS Number: 22620-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR