3-(3-Bromo-2-fluorophenyl)-2,4-dimethylpentan-3-ol ≥96%

Supplier: AOBChem USA

Warning

182351-1G 182351-500MG 182351-5G 182351-25G
77466-110EA 415.09 USD
77466-110 77478-854 77489-082 77499-536
3-(3-Bromo-2-fluorophenyl)-2,4-dimethylpentan-3-ol ≥96%
3-(3-Bromo-2-fluorophenyl)-2,4-dimethylpentan-3-ol

Caution: For research use only

Formula: C₁₃H₁₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD34857204
CAS Number: 1866622-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR