Butyl(2,5-difluorophenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

164558-1G 164558-500MG 164558-5G
77465-330EA 696.28 USD
77465-330 77478-092 77488-262
Butyl(2,5-difluorophenyl)sulfane ≥95%
Butyl(2,5-difluorophenyl)sulfane

Caution: For research use only

Formula: C₁₀H₁₂F₂S
Storage Temperature: Ambient
MDL Number: MFCD34850780
CAS Number: 1871718-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR