Tetramethylammonium hydroxide 10% in water

Supplier: TCI America
Danger

T0138-100ML T0138-25ML T0138-500ML
TCT0138-500MLEA 140.34 USD
TCT0138-500ML TCT0138-100ML TCT0138-25ML
Tetramethylammonium hydroxide 10% in water
Tetramethylammonium hydroxide

CAS Number: 75-59-2
MDL Number: MFCD00008280
Molecular Formula: C4H13NO
Molecular Weight: 91.15
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 37


Other Product Names / Synonyms: TMAH

Formula: C₄H₁₃NO
MW: 91.15 g/mol
Boiling Pt: 110 °C (1013 hPa)
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/44/62/28994462.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR