Tetrabutylammonium hydroxide 40% in water, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

MACRE224-80 MACRE224-80
MKE22480EA 280.42 USD
MKE22480 CAMKE22480
Tetrabutylammonium hydroxide 40% in water, Macron Fine Chemicals™
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Warm product to 30°C and shake prior to use.
Assay ((CH₃CH₂CH₂CH₂)₄NOH) 38 - 42 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR