Tricine ≥99%, white powder for electrophoresis
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid
, Tricine buffer
, N-(Tris(hydroxymethyl)methyl)glycine
Tricine was first prepared by Good for use as a buffer for chloroplast reactions. It is structurally similar to Tris, but is much less inhibitory at high concentrations. For ATP assays using firefly luciferase, tricine buffer at 25 mM was found to be the best of ten common buffers tested.
Tricine is used as buffer component for separation of low molecular weight peptides. Tricine can be used in cryopreservation medium for the preservation of tissues and organs. Cryopreservation depends on the physical and chemical characteristics of the preservation medium used. The pH values and pK values for tricine/DMSO mixtures has been reported down to -20 °C. Tricine has been found to be an efficient scavenger of hydroxyl radicals in a study of radiation-induced membrane damage. Tricine is typically the buffer of choice in SDS-PAGE systems when separating proteins in the range of 1 to 100 kDa.
A buffer may be prepared by titrating with sodium hydroxide to the desired pH, using about a half-equivalent of NaOH.
- Electrophoresis Grade
- Cell Culture Tested
- Presentation: White Powder
- pKa: 8.1 (25 °C)(Lit.)
- DpK/DT: -0.021(Lit.); Metal Binding Constants (log K) for 0.1 M solution at 20 °C: Mg2+: 1.2 Ca2+: 2.4 Mn2+: 2.7 Cu2+: 7.3(Lit.)
- Very soluble in water (25% w/v - clear, colorless solution).
- Store at Room Temperature (15-30 °C)
Formula:
(HOCH₂)₃CNHCH₂CO₂H MW: 179.17 g/mol Melting Pt: 179…189 °C (dec., lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00004277 CAS Number: 5704-04-1 EINECS: 227-193-6 Merck Index: 13,09724 |
Specification Test Results
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
OD 260 (0.1 M aq soln) | ≤0.1 |
OD 280 | ≤0.1 |
pH (1% aq soln) | 4.5-5.5 |
Moisture | ≤0.5% |
Heavy Metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...