Starch indicator Platner's
Supplier: Ricca Chemical
|
Danger
|
R8001000-500A
RC8001-16EA
130.74
USD
RC8001-16
Starch indicator Platner's
Starch
White, translucent liquid.
Formula:
(C₆H₁₀O₅)n MW: 162.14 g/mol Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0500 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00082026 CAS Number: 9005-25-8 |
Specification Test Results
Appearance | |
Suitability for Use | Colorless (Iodine absent) - Blue (Iodine present) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...