Sodium hydroxide 0.0833 N

Supplier: Ricca Chemical

Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

R7348000-20F
RC7348-5EA 279.51 USD
RC7348-5
Sodium hydroxide 0.0833 N
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.08330 ± 0.00005 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR