Potassium permanganate ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Permanganic acid potassium salt
Management of Change (MOC) category = N
BAKR3228-07
JT3228-7EA
1298.77
USD
JT3228-7
Potassium permanganate ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Potassium permanganate
Formula:
KMnO₄ MW: 158.03 g/mol Melting Pt: > 240 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011364 CAS Number: 7722-64-7 EINECS: 231-760-3 UN: 1490 ADR: 5.1,II Merck Index: 12,07824 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (KMnO₄) | ≥99.0 % |
Insoluble Matter | ≤0.2 % |
Chloride and Chlorate (as Cl) | ≤0.005 % |
Sulfate (SO₄) | ≤0.02 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...