Light Task Plus 3™ Polyurethane-Coated Cut-Resistant Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
NFD16G/8M NFD16G/9L NFD16G/10XL NFD16G/11XXL NFD16G/7S
89066-224CS 7672.24 USD
89066-224 89066-226 89066-228 89066-230 89066-222
Light Task Plus 3™ Polyurethane-Coated Cut-Resistant Gloves, Honeywell Safety
Gloves
Lightweight, cut level 3 gloves offer the perfect compromise between mechanical protection and wearer comfort.
  • Ideal for material handling and assembly work involving sharp objects
  • Seamless Dyneema® shell is stronger than steel and outperforms aramid fibers in cut and abrasion resistance
  • Tough polyurethane palm coating on palms is abrasion-and tear-resistant yet light enough to ensure dexterity and tactility
  • Glove back is uncoated for ventilation and comfort
  • Knit wrist
Ideal for material handling and assembly work involving sharp objects such as sheetMore Product Information