Potassium carbonate, anhydrous ≥99%, granular FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Carbonic acid potassium saltPotashCarbonic acid, dipotassium salt

Management of Change (MOC) category = N

BAKR3014-08 BAKR3014-08 BAKR3014-05 BAKR3014-05
JT3014-8EA 2981.46 USD
JT3014-8 JT3014-5 CAJT3014-5 CAJT3014-8
Potassium carbonate, anhydrous ≥99%, granular FCC, J.T.Baker®
Potassium carbonate

Granular.

Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891 °C
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://us.vwr.com/stibo/web/std.lang.all/05/98/16640598.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Assay (K₂CO₃) (dried basis) 99.0 - 100.5 %
FCC - Loss on Drying ≤1 %
FCC - Insoluble Substances Passes Test
FCC - Lead (Pb) ≤2 mg/kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR