D-(+)-Sucrose, Ultrapure

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: alpha-D-Glucopyranoside, β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, beta-D-Fructofuranosyl alpha-D-glucopyranoside, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, D-(+)-Saccharose, Sucrose, Sugar

04821713 04821721
IC821713EA 107.75 USD
IC821713 IC821721
D-(+)-Sucrose, Ultrapure
D(+)-Sucrose

Sucrose is a non-reducing disaccharide composed of glucose and fructose linked via their anomeric carbons. It is obtained commercially from sugarcane, sugar beet, and other plants and used extensively as a food and a sweetener. The mixture of glucose and fructose is known as "invert sugar."


  • DNAse and RNAse free
  • Concentrations of Heavy metals (as Pb): <5 ppm
  • Glucose: <0.1%


Sucrose is a simple carbohydrate used in density gradients and is hydrolyzed to glucose and fructose by dilute acids and by invertase, a yeast enzyme. Upon hydrolysis the optical rotation falls and is negative when the hydrolysis is complete. The mixture of glucose and fructose is known as "invert sugar." In a few cases, such as in riboflavin deficient diets, sucrose is preferred over corn starch due to its interactions in the gut to decrease intestinal synthesis of riboflavin, whereas corn starch has the opposite effect .It is used as a source of carbon in media and animal diet.8.5% (w/v) sucrose was used in dielectrophoretic field-flow fractionation (dFFF) that allows the rapid and convenient analysis of bulk cell suspensions (Sittisak Pui-ock,(2008),Dielectrophoretic Field-Flow Fractionation System for Detection of Aquatic Toxicants,Analytical Chemistry,80(20),pages: 7727-7734).


Soluble in water (1 g/0.5 mL at 25°C; 1 g/0.2 mL boiling water), ethanol (1 g/170 mL) or methanol (1 g/100 mL); moderately soluble in glycerol or pyridine.

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms Beet sugar; Cane sugar; b-D-Fructofuranosyl-a-D-glucopyranoside; D-(+)-Saccharose; Sugar; a-D-Glucopyranosyl-b-D-fructofuranoside
Form White crystalline powder
Storage Store at Room Temperature (15-30 °C)
Heavy Metals ≤0.0005% (Pb)
Optical Rotation +66.0°±5 (H2O, NH4OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR