Sodium dodecyl sulfate (SDS) ≥99%, fine white powder, Ultrapure
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
Sodium dodecyl sulfate
, Lauryl sulfate sodium salt
, Sodium lauryl sulfate
, Dodecyl sulfate sodium salt
, Dodecyl sulphuric acid sodium salt
, Lauryl sulphate sodium salt
, Sodium lauryl sulphate
, Dodecyl sulphate sodium salt
, Dodecyl sulfuric acid sodium salt
SDS is an anionic detergent and wetting agent that is effective in both acid and alkaline solutions.
Used to solubilize and denature proteins for denaturing-PAGE. Most proteins bind SDS in a ratio of 1.4 g SDS per gram of protein. The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS. The charge to mass ratio is essentially the same for each protein and will migrate in the gel based only on their size.
- Ultra Pure Grade
- Suitable for Electrophoresis
- Presentation: Fine White Powder
- Aggregation Number: 62(Lit.)
- Critical Micelle Concentration (CMC): 7-10 mM (Lit.)
- Soluble in water (200 mg/mL - clear, faint yellow solution), and ethanol (0.1g/10 mL).
- Store at Room temperature(15-30 °C)
Formula:
CH₃(CH₂)₁₁OSO₃Na MW: 288.38 g/mol Melting Pt: 180…210 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,III Merck Index: 13,08710 |
Specification Test Results
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
Melting Point | 180-210°C |
OD260 (1% aq soln) | ≤0.1 |
OD280 (1% aq soln) | ≤0.1 |
pH (1% aq soln) | 6-7 |
Heavy Metals (as Pb) | ≤20 ppm |
Phosphate | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...