Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PacOrange)

Supplier: AAT BIOQUEST INC
100411K0 100411K1
77022-534EA 244.24 USD
77022-534 77021-168
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PacOrange)
Antibodies
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: PacOrange
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR