Power Strips, Tripp Lite
Supplier: Tripp Lite
PS3612 PS7224-20
89079-144EA 115.65 USD
89079-144 89079-146
Power Strips, Tripp Lite
Electrical Sockets
Power strips provide 120V, 50/60Hz AC power distribution with a circuit breaker in a mountable aluminum housing, ideal for horizontal or vertical rackmounts or raceways in office, workbench, industrial, networking, and telecom applications
Lighted power switch with locking transparent cover prevents accidental shutoff. Color: silver with black receptacles and power cord.
Certifications: UL 1363 (power tap) and cUL listed.
Ordering information: Manufacturer's lifetime warranty. Supplied with mounting hardware for vertical or horizontal mounting.