Sodium acetate, anhydrous ≥99.0%, GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: Acetic acid sodium salt

CA1.06268.2500
EMD-SX0265-3EA 701.57 USD
EMD-SX0265-3
Sodium acetate, anhydrous ≥99.0%, GR ACS, Supelco®
Sodium acetate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Assay 99.0% min
Calcium (Ca) 0.005% max
Chloride (Cl) 0.002% max
Heavy metals (as Pb) 0.001% max
Insoluble matter 0.01% max
Iron (Fe) 0.001% max
Loss on drying (120°C) 1.0% max
Magnesium (Mg) 0.002% max
pH of a 5% solution at 25°C 7.0-9.2
Phosphate (PO4) 0.001% max
Sulfate (SO4) 0.003% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR