Anti-CD19 Mouse Monoclonal Antibody (PE/iFluor® 594) [clone: HIB19]

Supplier: AAT BIOQUEST INC
101921Y0 101921Y1 101921Y2
77023-352EA 210.7 USD
77023-352 77024-130 77024-366
Anti-CD19 Mouse Monoclonal Antibody (PE/iFluor® 594) [clone: HIB19]
Antibodies
HIB19 is an anti-human monoclonal antibody that recognizes the CD19 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR