Anti-CD16a Mouse Monoclonal Antibody (iFluor® 860) [clone: 3G8]

Supplier: AAT BIOQUEST INC
101610R1 101610R0
77023-106EA 915.16 USD
77023-106 77021-648
Anti-CD16a Mouse Monoclonal Antibody (iFluor® 860) [clone: 3G8]
Antibodies
The 3G8 monoclonal antibody recognizes human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: iFluor® 860
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR