Anti-CD8a Mouse Monoclonal Antibody (mFluor™ Violet) [clone: SK1]

Supplier: AAT BIOQUEST INC
10081110 10081111
77021-650EA 244.24 USD
77021-650 77024-054
Anti-CD8a Mouse Monoclonal Antibody (mFluor™ Violet) [clone: SK1]
Antibodies
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: mFluor™ Violet
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR