Anti-CD5 Mouse Monoclonal Antibody (PE/iFluor® 700) [clone: UCHT2]

Supplier: AAT BIOQUEST INC
100521X0 100521X1 100521X2
77024-258EA 210.7 USD
77024-258 77022-264 77023-228
Anti-CD5 Mouse Monoclonal Antibody (PE/iFluor® 700) [clone: UCHT2]
Antibodies
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: PE/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR