Sunlite® LED Growing Centers
Supplier: GARDENERS SUPPLY COMPANY
8595555 8595556 8595557 8595552 8595553 8595554
470355-168EA 291.65 USD
470355-168 470355-176 470355-188 470355-190 470355-170 470355-180
Sunlite® LED Growing Centers
Gardening Supplies
Find the perfect grow center for your class.
  • Powder-coated aluminum frame, recycled plastic, aluminum, polycarbonate
  • Full spectrum LED bulbs included
  • Adjustable light height
  • Compact and full size versions available
These SunLite Gardens features revolutionary LED lighting technology, creating the new gold standard for seed starting and indoor gardening. You'll be amazed at how well your seedlings, houseplants, orchids, and other indoor plants grow as they bask in the glow of these specialized LEDs. The compact frames hold one light fixture per tier that can easily be raised and lowered to the perfect height for plants. Full size frames hold 2 bulbs per tier.