Alaris IV Pump Modules
Supplier: GUMBO MEDICAL
A8015P A8100P A8110P A8120P A815810 A8300P
470354-742EA 1307.69 USD
470354-742 470354-724 470354-732 470354-728 470354-726 470354-718
Alaris IV Pump Modules
Infusions
Perfect for your Sim center.
  • Available as a bundle or individual components
  • Refurbished
The Alaris Medley Combo is a 2 module combination that allows users to have an out of the box solution for a patient care system. This system includes 1 each Alaris Medley 8015 PC Module, and 1 each Alaris Medley 8100 IV Pump Module.
The BD/Alaris Medley Point of Care System with Guardrails Suite MX software is a Modular smart IV medication safety system that can help protect every infusion to guard you and your patients from medication errors. The Alaris Medley Point of Care System combines a single point-of-care unit featuring Guardrails Suite MX safety software with IV/LVP, PCA Syringe, Syringe, respiratory monitoring (SpO₂ and EtCO₂), and even barcoding capabilities.
Each Alaris Medley Combo is Guardrail compatible, and Educational customers have the option of having an Educational Training Dataset added for training purposes. A single 8015 PC Module can support up to four Infusion Modules with a single power cord, and features a large, easy to read color display and back-lit keypad for programming values. Compared to the Medley 8000 PC module, the newer 8015 Module offers similar functionality with a brighter, color display and Wireless communications capability. The Alaris Medley 8100 IV pump modules are devices that can help protect every infusion type, and are designed for the delivery of fluids, medications, blood, and blood products using continuous or intermittent delivery for adult, pediatric, or neonatal patients. The Alaris Medley system is the most flexible patient care available today.
Ordering information: Additional features and models may be available. Please contact your sales representative or customer service for information.
Caution: This item is refurbished and is intended for educational and training use only.