Nitrile Finger Cots, Honeywell Safety
Supplier: Honeywell Safety Products
135LNR/L-H5 115LNR/S 125LNR/M-H5 145LNR/XL
11000-998CS 3229.95 USD
11000-998 11001-002 11001-000 11001-004
Nitrile Finger Cots, Honeywell Safety
Finger Cots Nitrile Finger Cots
Multiple-washed, powder-free finger cots are very low in particulates
Inherently static-dissipative, these finger cots have an average surface resistivity of 1011ohms/sq. A unique low modulus nitrile formulation offers the stretch and comfort of natural rubber without allergy-causing latex proteins. Each finger cot is rolled for easy donning. Color: white.